ENA638597661 Enamine

5-(1H-1,3-benzodiazol-2-yl)pyridin-3-amine

  • MDL number MFCD11107321
  • Cas Number : 1082159-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342380
Availability details for
Quantity
  • Availability
  • Add to Cart