ENA638597675 Enamine

tert-butyl N-(4-amino-1-phenylbutan-2-yl)carbamate

  • MDL number MFCD24532822
  • Cas Number : 159025-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342418
Availability details for
Quantity
  • Availability
  • Add to Cart