ENA638597682 Enamine

3-phenyl-1,8-naphthyridin-2-amine

  • MDL number MFCD11588086
  • Cas Number : 5174-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342434
Availability details for
Quantity
  • Availability
  • Add to Cart