ENA638597764 Enamine

2-(chloromethyl)-5-(2-nitrophenyl)-1,3,4-oxadiazole

  • MDL number MFCD00970479
  • Cas Number : 63002-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342646
Availability details for
Quantity
  • Availability
  • Add to Cart