ENA638597765 Enamine

N-(but-2-yn-1-yl)furan-2-carboxamide

  • MDL number MFCD24618497
  • Cas Number : 1333470-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342647
Availability details for
Quantity
  • Availability
  • Add to Cart