ENA638597859 Enamine

2-acetamidooctanoic acid

  • MDL number MFCD18862917
  • Cas Number : 80782-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342146
Availability details for
Quantity
  • Availability
  • Add to Cart