ENA638597875 Enamine

2-(2-aminoethyl)-1,3-thiazole-4-carboxylic acid dihydrochloride

  • MDL number MFCD16295317
  • Cas Number : 1215336-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343126
Availability details for
Quantity
  • Availability
  • Add to Cart