ENA638597879 Enamine

6-chloro-N-(2,2-difluoroethyl)-2-methylpyrimidin-4-amine

  • MDL number MFCD23728377
  • Cas Number : 1552918-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342178
Availability details for
Quantity
  • Availability
  • Add to Cart