ENA638597910 Enamine

2-(1-methylcyclopropyl)-1-(pyridin-4-yl)propan-2-ol

  • MDL number MFCD23870701
  • Cas Number : 1528577-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343226
Availability details for
Quantity
  • Availability
  • Add to Cart