ENA638597915 Enamine

1,3-dimethylcyclohexan-1-ol

  • MDL number MFCD12067491
  • Cas Number : 55539-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343231
Availability details for
Quantity
  • Availability
  • Add to Cart