ENA638597916 Enamine

3-{5-methyl-3H-imidazo[4,5-b]pyridin-2-yl}aniline

  • MDL number MFCD25074621
  • Cas Number : 1533523-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343234
Availability details for
Quantity
  • Availability
  • Add to Cart