ENA638597922 Enamine

4-cyclohexyl-1,3-thiazole-2-thiol

  • MDL number MFCD20293754
  • Cas Number : 2180-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343241
Availability details for
Quantity
  • Availability
  • Add to Cart