ENA638597935 Enamine

1,5-dichloro-3-fluoro-2-isothiocyanatobenzene

  • MDL number MFCD28166241
  • Cas Number : 1823548-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343260

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P272,P280,P285,P301+P310,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P304+P341,P305+P351+P338,P310,P330,P333+P313,P342+P311,P363,P405,P501
hcodes T,C
MSDS https://www.enaminestore.com/catalog/EN300-343260
pictograms GHS05,GHS06,GHS08
hstatements H301,H314,H317,H334
Availability details for
Quantity
  • Availability
  • Add to Cart