ENA638597944 Enamine

1-benzyl-2,3,4,5-tetrahydro-1H-1-benzazepin-5-one

  • MDL number MFCD30535725
  • Cas Number : 890839-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343285
Availability details for
Quantity
  • Availability
  • Add to Cart