ENA638597957 Enamine

1-benzyl-4-(3,4-dichlorophenyl)-1H-imidazol-5-amine

  • MDL number MFCD30254053
  • Cas Number : 1455381-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343333
Availability details for
Quantity
  • Availability
  • Add to Cart