ENA638598503 Enamine

acetic acid; tert-butyl N-(1-carbamimidoylcyclobutyl)carbamate

  • MDL number MFCD30535883
  • Cas Number : 2060042-17-1
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CH3COOH
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-344554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-344554
Availability details for
Quantity
  • Availability
  • Add to Cart