ENA638598601 Enamine

3-bromo-4-(chloromethyl)-1-methyl-1H-pyrazole

  • MDL number MFCD30536015
  • Cas Number : 2060043-12-9
  • Molecular Weight : 209.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-345522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-345522
Availability details for
Quantity
  • Availability
  • Add to Cart