ENA638598700 Enamine

[1-(4-amino-2-bromophenyl)pyrrolidin-3-yl]methanol

  • MDL number MFCD26059785
  • Cas Number : 1565386-11-9
  • Molecular Weight : 271.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-344480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-344480
Availability details for
Quantity
  • Availability
  • Add to Cart