ENA638598932 Enamine

[(4-chloropyridin-2-yl)methyl]dimethylamine

  • MDL number MFCD21833563
  • Cas Number : 315493-84-6
  • Molecular Weight : 170.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-345139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-345139
Availability details for
Quantity
  • Availability
  • Add to Cart