ENA638599119 Enamine

1-(2-fluoroethyl)-1H-pyrazol-3-amine

  • MDL number MFCD23130701
  • Cas Number : 1258863-98-7
  • Molecular Weight : 129.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-345687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-345687
Availability details for
Quantity
  • Availability
  • Add to Cart