ENA638599287 Enamine

8-chloro-5,6,7,8-tetrahydroquinoline hydrochloride

  • MDL number MFCD18803322
  • Cas Number : 114432-00-7
  • Molecular Weight : 204.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-358863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-358863
Availability details for
Quantity
  • Availability
  • Add to Cart