ENA715369377 Enamine

3-methyl-2-phenylmorpholin-2-ol hydrochloride

  • MDL number MFCD28023806

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178659
Availability details for
Quantity
  • Availability
  • Add to Cart