ENA715369449 Enamine

1-(3-methylphenyl)propan-2-one

  • MDL number MFCD00210466
  • Cas Number : 18826-61-4
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-225303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225303
Availability details for
Quantity
  • Availability
  • Add to Cart