ENA715369628 Enamine

2-(aminomethyl)-2-methylbutanoic acid hydrochloride

  • MDL number MFCD22628372
  • Cas Number : 1485425-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118992

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118992
Availability details for
Quantity
  • Availability
  • Add to Cart