ENA769494988 Enamine

[4-(dimethylamino)-6-hydrazinylpyrimidin-2-yl]methanol

  • MDL number MFCD30537765
  • Cas Number : 2095410-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-345132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-345132
Availability details for
Quantity
  • Availability
  • Add to Cart