ENA769500318 Enamine

1-[(tert-butoxy)carbonyl]-6-fluoro-2,3-dihydro-1H-indole-2-carboxylic acid

  • MDL number MFCD30539051
  • Cas Number : 2137506-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-361458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-361458
Availability details for
Quantity
  • Availability
  • Add to Cart