ENA769500734 Enamine

6-bromoquinolin-3-amine

  • MDL number MFCD09260429
  • Cas Number : 930570-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-361747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-361747
Availability details for
Quantity
  • Availability
  • Add to Cart