ENA769500855 Enamine

4,8-dichloro-3-nitroquinoline

  • MDL number MFCD28955313
  • Cas Number : 39487-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-361534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-361534
Availability details for
Quantity
  • Availability
  • Add to Cart