ENA769503823 Enamine

3-[(1,3-thiazol-2-yl)methyl]piperidine

  • MDL number MFCD09891854
  • Cas Number : 1017215-27-8
  • Molecular Weight : 182.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-363108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-363108
Availability details for
Quantity
  • Availability
  • Add to Cart