ENA769505466 Enamine

2-chloro-4-[(piperazin-1-yl)methyl]phenol dihydrochloride

  • MDL number MFCD30539817
  • Cas Number : 2089257-94-1
  • Molecular Weight : 299.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-365529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-365529
Availability details for
Quantity
  • Availability
  • Add to Cart