ENA769505744 Enamine

tert-butyl 4-amino-4-(2-methoxyethyl)piperidine-1-carboxylate

  • MDL number MFCD27995813
  • Cas Number : 1713163-25-7
  • Molecular Weight : 258.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-364721

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-364721
Availability details for
Quantity
  • Availability
  • Add to Cart