ENA769505991 Enamine

3-bromo-5-fluoro-1H-indole

  • MDL number MFCD12828703
  • Cas Number : 1186663-46-6
  • Molecular Weight : 214.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-365156

Safety Information

signalword Danger
pcodes P210,P230,P234,P240,P250,P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P363,P370+P380,P372,P373,P401,P501
MSDS https://www.enaminestore.com/catalog/EN300-365156
pictograms GHS07
hstatements H205,H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart