ENA769507687 Enamine

tert-butyl N-[(1S)-1-(2-hydroxyphenyl)ethyl]carbamate

  • MDL number MFCD30539871
  • Cas Number : 2089246-47-7
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-365946

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-365946
Availability details for
Quantity
  • Availability
  • Add to Cart