ENA769508066 Enamine

2-(aminomethyl)-6-fluorophenol

  • MDL number MFCD07786745
  • Cas Number : 887583-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366174
Availability details for
Quantity
  • Availability
  • Add to Cart