ENA769508165 Enamine

ethyl 2-(4-cyclohexyl-1,3-thiazol-2-yl)acetate

  • MDL number MFCD12567076
  • Cas Number : 1152845-40-3
  • Molecular Weight : 253.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-366810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366810
Availability details for
Quantity
  • Availability
  • Add to Cart