ENA769508203 Enamine

{[4-bromo-2-(3-chloro-4-fluorophenoxy)phenyl]methyl}(methyl)amine

  • MDL number MFCD19197447
  • Cas Number : 1275536-49-6
  • Molecular Weight : 344.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-370634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-370634
Availability details for
Quantity
  • Availability
  • Add to Cart