ENA769508327 Enamine

2-(4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)acetic acid

  • MDL number MFCD26938129
  • Cas Number : 1518990-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-370960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-370960
Availability details for
Quantity
  • Availability
  • Add to Cart