ENA769508378 Enamine

ethyl (3R)-3-amino-3-(2-chlorophenyl)propanoate hydrochloride

  • MDL number MFCD30375794
  • Cas Number : 2089245-85-0
  • Molecular Weight : 264.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-365882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-365882
Availability details for
Quantity
  • Availability
  • Add to Cart