ENA769508394 Enamine

3-methyl-1-phenylbutan-2-amine

  • MDL number MFCD11934510
  • Cas Number : 46114-16-3
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-371176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-371176
Availability details for
Quantity
  • Availability
  • Add to Cart