ENA769508510 Enamine

2-(2,5-dimethyl-1-phenyl-1H-pyrrol-3-yl)acetonitrile

  • MDL number MFCD07345669
  • Cas Number : 100869-86-1
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-371882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-371882
Availability details for
Quantity
  • Availability
  • Add to Cart