ENA769508553 Enamine

2-(oxan-2-yl)propan-1-ol

  • MDL number MFCD28646105
  • Cas Number : 1782898-92-3
  • Molecular Weight : 144.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-372042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-372042
Availability details for
Quantity
  • Availability
  • Add to Cart