ENA769508555 Enamine

2,4-dimethoxypyridin-3-amine

  • MDL number MFCD18255875
  • Cas Number : 217096-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-372046

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-372046
Availability details for
Quantity
  • Availability
  • Add to Cart