ENA769508629 Enamine

2-(ethoxymethyl)-4-hydrazinyl-6-methylpyrimidine

  • MDL number MFCD21297516
  • Cas Number : 1496071-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-373411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-373411
Availability details for
Quantity
  • Availability
  • Add to Cart