ENA769517721 Enamine

5-{1-[(tert-butoxy)carbonyl]pyrrolidin-2-yl}-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD30724003
  • Cas Number : 1694220-62-6
  • Molecular Weight : 282.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-374316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-374316
Availability details for
Quantity
  • Availability
  • Add to Cart