ENA769517805 Enamine

2-(dimethyl-1,3-thiazol-2-yl)piperidine

  • MDL number MFCD16789587
  • Cas Number : 1248899-34-4
  • Molecular Weight : 196.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62603
Availability details for
Quantity
  • Availability
  • Add to Cart