ENA769517861 Enamine

2-[3-(hydroxyimino)cyclopentyl]-2,3-dihydro-1H-isoindole-1,3-dione

  • Cas Number : 1937228-69-7
  • Molecular Weight : 244.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-97885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97885
Availability details for
Quantity
  • Availability
  • Add to Cart