ENA946831477 Enamine

3-amino-3-(2,6-difluorophenyl)propanoic acid hydrochloride

  • MDL number MFCD30730419
  • Cas Number : 2095410-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-295794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-295794
Availability details for
Quantity
  • Availability
  • Add to Cart