ENA946831486 Enamine

2-chloro-N1-(4,5-dihydro-1H-imidazol-2-yl)benzene-1,4-diamine dihydrochloride

  • MDL number MFCD30730427
  • Cas Number : 21571-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-310835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-310835
Availability details for
Quantity
  • Availability
  • Add to Cart