ENA946831487 Enamine

3-(chloromethyl)-4-methyl-1H-pyrazole hydrochloride

  • MDL number MFCD30730429
  • Cas Number : 2095410-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-311041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-311041
Availability details for
Quantity
  • Availability
  • Add to Cart