ENA946831489 Enamine

1-benzyl-2-(3,3-dimethylazetidin-2-yl)-1H-imidazole

  • MDL number MFCD30673867
  • Cas Number : 2095410-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-312108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-312108
Availability details for
Quantity
  • Availability
  • Add to Cart